N,N-Diethyl-m-toluamide 98%
:134-62-3
:
:C₁₂H₁₇NO
:MFCD00009046
:191.27 g/mol
:13,02876
:111 °C (1 mmHg)
:-45 °C
:0.9955 g/cm³
:155 °C
:205-149-7
:GHS07
:Ambient
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.5200 to 1.5240 (25°C, 589 nm)
- Water:≤0.3 %
- GC:≥97.5 %
- Color scale:≤100 APHA
Frequently Bought Together