Order Entry
France
ContactUsLinkComponent
Pyridoxine hydrochloride
Pyridoxine hydrochloride
Catalog # MOLE13101713-25G
Supplier:  Molekula
CAS Number:  
Pyridoxine hydrochloride
Catalog # MOLE13101713-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :205.64 g/mol
Merck Index:13,08072
Melting Pt:214…215 °C
Storage Temperature:Ambient
EINECS:200-386-2
CAS Number:58-56-0
Formula:C₈H₁₂ClNO₃
MDL Number:MFCD00012807