Gentle Cell Dissociation Reagent
Supplier: STEMCELL Technologies
Certificates
About this item
cGMP, enzyme-free cell dissociation reagent.
- Obtain quality cultures with this gentle, chemically-defined, enzyme-free, GMP solution
- Achieve high expansion of human ES/iPS cells during routine culture
- Simplify cell passaging with straightforward, room temperature passaging protocols
Use Gentle Cell Dissociation Reagent (GCDR) to dissociate human embryonic stem (ES) cells or human induced pluripotent stem (iPS) cells into cell aggregates for routine passaging, or into a single-cell suspension. This enzyme-free and chemically-defined reagent ensures high expansion of human ES and iPS cells during routine culture. It is also suitable for the isolation of intestinal crypts to establish intestinal organoids, and for breaking up Corning® Matrigel® domes when passaging organoid cultures.
GCDR is now also manufactured following relevant cGMPs under a certified quality management system to ensure the highest quality and consistency for reproducible results.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
- Species:HumanMouse