Order Entry
France
ContactUsLinkComponent
Collagenase Type I (0,25%)
Collagenase Type I (0,25%)
Catalog # STMC07902
CAS Number:  
undefined
Collagenase Type I (0,25%)
Catalog # STMC07902
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Species:
    Non-human primate
    Human
    Rat
    Mouse
  • Description:
    Collagenase type I (0,25%)

Specifications

About this item

Cell dissociation reagent.

  • Optimised for the preparation of cell suspensions, dissociation of mouse embryoid bodies (EBs), and solubilisation of collagen gels

Digest native collagen fibrils in connective tissues with Collagenase Type I (0,25%). This Collagenase Type I solution has been optimised for the preparation of cell suspensions, dissociation of mouse embryoid bodies (EBs), and solubilisation of collagen gels. Sourced from Clostridium histolyticum, this enzymatic preparation contains the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin.

Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').