Drawer Racks, Stainless Steel
Supplier: RATIOLAB
New Product
Certificates
About this item
Stainless steel racks for storing the cryo boxes in freezer cabinets and chests.
- Cabinet slide-ins without pull-out stop
- Easy-to-clean polished surface
- Robust design
Racks for for all cryo boxes format 130×130; 133×133 and 136×136 mm.
Frequently Bought Together