5-Nitro-2-(3-phenylpropylamino)benzoic acid 98%
:107254-86-4
:
:C₁₆H₁₆N₂O₄
:MFCD00153851
:300.314 g/mol
:Freezer
- Appearance:yellow powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals