Order Entry
France
ContactUsLinkComponent
Fumaric acid monomethyl ester 97%
Fumaric acid monomethyl ester 97%
New Product
Certificates
Fumaric acid monomethyl ester 97%
New Product

Specification Test Results

  • Appearance:
    white to almost white powder oe crystals
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥96.0 %
  • Melting point:
    145°C to 150°C

Specification Test Results

Frequently Bought Together