0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
France
ContactUsLinkComponent
Wright's stain
Wright's stain for microscopical staining
Wright's stain for microscopical staining
MOLE57163864-25G
:
Molekula
:
Wright's stain for microscopical staining
MOLE57163864-25G
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
0.8 g/cm³ (20 °C)
:
273-541-5
:
GHS07
:
Ambient
:
68988-92-1
:
51938-72-8
:
MFCD00082143