Saisie de commande
France
Lien pour commanderContactUsLinkComponent
Formaldéhyde 4%

Formaldéhyde 4%

Health
Irritant
Supplier:   PROLABO CMP
Formaldéhyde 4%
Supplier:  PROLABO CMP
Certificates
Formaldéhyde 4%

2 Options Available Below

Product Details & Documents

(10% Formaline solution)

Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Storage Temperature:Température ambiante
  • EINECS:200-001-8
Product Family Options

Product Information

  • Type de conditionnement
    Cdt
    Availability
    Price
  • 10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 217,00 €
    217,00 €
    1 * 10 L
       217,00 €

  • Flacon Plastique
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 239,00 €
    239,00 €
    1 * 10 L
       239,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.