Ethanol absolute analytical reagent, reference substance for gas chromatography, Supelco®
Glass bottle, 5 mL
Supplier: Merck
Ethanol absolute analytical reagent, reference substance for gas chromatography, Supelco®
Catalog # 02483-5ML
Supplier: Merck
CAS Number: 64-17-5
Jump to:
Product Details & Documents
Synonyms: Ethyl alcohol
- CAS Number:64-17-5
- Formula:C₂H₅OH
- MDL Number:MFCD00003568
- Molecular Weight:46.07 g/mol
- Merck Index:12,03806
- Boiling Pt:78,3 °C (1013 hPa)
- Melting Pt:–117 °C
- Density:0,7895 g/cm³ (20 °C)
- Flash Pt:12 °C
- Storage Temperature:Ambient
- EINECS:200-578-6
- UN:1170
- ADR:3,II
- REACH:01-2119457610-43
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle
- Pk:5 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



