Diisopropylcarbodiimide
Catalog # MOLEM79451433
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:126.2 g/mol
Boiling Pt:145 °C (1013 hPa)
Density:0.81 g/cm³ (20 °C)
Flash Pt:33 °C
EINECS:211-743-7
UN:2929
ADR:6.1,II
CAS Number:693-13-0
Formula:C₇H₁₄N₂
MDL Number:MFCD00065689