Order Entry
France
ContactUsLinkComponent
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Catalog # PIER28300
CAS Number:  
undefined
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Catalog # PIER28300
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :614.89 g/mol
Merck Index:13,02052
Melting Pt:116 °C
Storage Temperature:Refrigerator
CAS Number:75621-03-3
Formula:C₃₂H₅₈N₂O₇S
MDL Number:MFCD00081083