Order Entry
France
Orders LinkContactUsLinkComponent
 

Column Coupler

Column coupler, 100×0,063 mm, Stainless steel, 0,70 mm
  MAIS70.10.070
 :   DR MAISCH
Column Coupler
  MAIS70.10.070
 :  DR MAISCH
Column Coupler

Column coupler, 100×0,063 mm, Stainless steel, 0,70 mm

 
 
   
1 * 1 Pce
 
 
  • Ø int.:
    0,70 mm
  • Description:
    Column coupler, 100×0,063 mm
  • Material:
    Stainless steel
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.