Supelco® Cerilliant® Organic Reference Standard, 5-Hydroxyindole-2,4,6,7-d4-3-acetic-2,2-d2 acid (5-HIAA-D6) solution
H-152-1ML
: Merck
:
- Concentration:100 µg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- Description:5-Hydroxyindole-2,4,6,7-d4-3-acetic-2,2-d2 acid (5-HIAA-D6) solution
- Environmentally Preferable:
- Storage conditions:–70 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:DICE
- Molecular weight:197,22
- Flash point:9,7 °C - Closed cup - Not applicable
- Pk Info:Ampule of 1 ml
: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311