Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
XFD647 AML-NHS ester
XFD647 AML-NHS ester, ReadiCleave™
XFD647 AML-NHS ester, ReadiCleave™
XFD647 AML-NHS ester, ReadiCleave™
Catalog # AATB7012
Supplier:
AAT Bioquest
New Product
CAS Number:
Expand image
XFD647 AML-NHS ester, ReadiCleave™
Catalog # AATB7012
Supplier:
AAT Bioquest
New Product
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
1609 g/mol
Product Details