Order Entry
France
Orders LinkContactUsLinkComponent
 

Protonex™ NIR 770 NHS ester

1 mg
Catalog # AATB21221
Supplier:   AAT Bioquest
Protonex™ NIR 770 NHS ester
Catalog # AATB21221
Supplier:  AAT Bioquest
CAS Number:  
Protonex™ NIR 770 NHS ester

1 mg

CAS Number:  
Price
544,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

Protonex™ NIR 770 NHS ester is the only cell-permeable acidotropic pH dye commercially available for developing NIR pH probes.

  • Stable for 12 months

Quality tested in-house

 : For Research Use Only

  • Molecular Weight:535.64 g/mol
Specifications

Specifications

  • Pk:
    1 mg

Specifications