Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2’,3’-cGAMP-DBCO
2’,3’-cGAMP-DBCO conjugate
2’,3’-cGAMP-DBCO conjugate
2’,3’-cGAMP-DBCO conjugate
Catalog # AATB20330
Fournisseur:
AAT Bioquest
New Product
Numéro CAS:
Expand image
2’,3’-cGAMP-DBCO conjugate
Catalog # AATB20330
Fournisseur:
AAT Bioquest
New Product
Numéro CAS:
Spécifications
Pk:
100 µG
Seq:
1286
Spécifications
À propos de cet article
Product Details
:
:
1539.59 g/mol
Product Details