Anti-Bombesin Receptor 3 Rabbit Polyclonal Antibody
ANTIA100011-100
New Product
- Antibody type:Primary
- Antigen name:BB3
- Antigen symbol:Bombesin Receptor 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P32247
- Antigen synonyms:BLP receptor subtype 3|BRS3_HUMAN|Bombesin Receptor 3|Bombesin receptor subtype 3|Bombesin receptor subtype-3|BRS-3|Bombesin like receptor 3|BRS3|BRS 3
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:44 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human BRS3 (amino acids 161 - 210)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to BRS3 for WB, IF and ELISA with samples derived from human.
- Validated Applications: WB, IF, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:40000
Type: Primary
Antigen: Bombesin Receptor 3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human