Anti-TCF3/E2A Rabbit Polyclonal Antibody
Catalog # ANTIA94108-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:AGM8
- Antigen symbol:TCF3/E2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# P15923
- Antigen synonyms:E12|Immunoglobulin enhancer-binding factor E12/E47|Helix loop helix protein HE47|bHLHb21|Class B basic helix-loop-helix protein 21|ITF1|E2A|E2A immunoglobulin enhancer binding factors E12/E47|Immunoglobulin transcription factor 1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:67 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human E2A around the phosphorylation site of Thr355 (amino acids 321-370)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to E2A (phospho Thr355) for IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: IHC, ELISA
- Recommended dilutions: IHC: 1:50 to 1:100, ELISA: 1:1000
Type: Primary
Antigen: TCF3/E2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat