Anti-MEF2D Mouse Monoclonal Antibody [clone: PCRP-MEF2D-3A4]
ANTIA277705-100
New Product
- Antibody type:Primary
- Antigen name:DKFZp686I1536
- Antigen symbol:MEF2D
- Clonality:Monoclonal
- Clone:PCRP-MEF2D-3A4
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q14814
- Antigen synonyms:MEF2D_HUMAN|Myocyte specific enhancer factor 2D|Myocyte specific enhancer factor 2, polypeptide D|MADS box transcription factor 2 polypeptide D|Myocyte-specific enhancer factor 2D|Myocyte enhancer factor 2D|MEF2D
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:55,94 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human MEF2D protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [PCRP-MEF2D-3A4] antibody to MEF2D for IP, Flow Cytometry, IF and IHC-P with samples derived from human.
- Validated Applications: IP, Flow Cytometry, IF, IHC-P
- Recommended Dilutions: IP: 1 to 2µg / 100 to 500µg proteins, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: MEF2D
Clonality: Monoclonal
Clone: PCRP-MEF2D-3A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human