Anti-CD63 Rabbit Recombinant Antibody [clone: LAMP3/2990R]
ANTIA253935-100
New Product
- Antibody type:Primary
- Antigen name:CD 63
- Antigen symbol:CD63
- Clonality:Recombinant
- Clone:LAMP3/2990R
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P08962
- Antigen synonyms:CD63_HUMAN|gp55|CD63|CD63 molecule|Granulophysin|CD63 antigen (melanoma 1 antigen)|CD63 antigen|LAMP 3|CD63 antigen melanoma 1 antigen
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:26 kDa (core protein)/30 - 60 kDa (glycosylated)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human CD63 protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [LAMP3/2990R] antibody to CD63 for Flow Cytometry and IHC-P with samples derived from human.
- Validated Applications: Flow Cytometry, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD63
Clonality: Recombinant
Clone: LAMP3/2990R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human