Anti-FOXL1 Mouse Monoclonal Antibody [Clone: PCRP-FOXL1-1F8]
Catalog # ANTIA278193-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:FKH 6
- Antigen symbol:FOXL1
- Clonality:Monoclonal
- Clone:PCRP-FOXL1-1F8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q12951
- Antigen synonyms:Forkhead box protein L1|Forkhead box L1|Forkhead related protein FKHL11|Forkhead related transcription factor 7|Forkhead (Drosophila) like 11|FKHL11|Forkhead-related protein FKHL11|FKHL 11|FKH6
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:36-41 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 104-258, of human FOXL1 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [PCRP-FOXL1-1F8] antibody to FOXL1 for IP, Flow Cytometry, IF, WB and IHC-P with samples derived from human.
- Validated Applications: IP, Flow Cytometry, IF, WB, IHC-P
- Recommended Dilutions: IP: 1 to 2µg / 100 to 500µg proteins, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: FOXL1
Clonality: Monoclonal
Clone: PCRP-FOXL1-1F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human