Anti-MASH1/Achaete-scute homolog 1 Rabbit Polyclonal Antibody
ANTIA95934-100
New Product
- Antibody type:Primary
- Antigen name:Achaete scute complex homolog 1
- Antigen symbol:MASH1/Achaete-scute homolog 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P50553
- Antigen synonyms:ascl1|Achaete scute complex homologue 1|Achaete scute protein|Achaete scute complex like 1|Achaete scute complex homolog like 1|Ascl 1|Achaete scute family bHLH transcription factor 1|Achaete-scute homolog 1|Achaete scute complex homologue like 1
- Storage buffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:25 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human ASCL1 (amino acids 109-158)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to ASCL1 for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, IHC, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: MASH1/Achaete-scute homolog 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat