Anti-Serum Response Factor SRF Rabbit Polyclonal Antibody
Catalog # ANTIA94938-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:c fos serum response element binding factor
- Antigen symbol:Serum Response Factor SRF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Form:Liquid
- Gene ID:UniprotID# P11831
- Antigen synonyms:ELK3|MCM 1|c fos serum response element binding transcription factor|ERP|Serum response factor|OTTHUMP00000039820|Serum Response Factor SRF|MCM1|SAP2
- Storage buffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:51 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human SRF around the phosphorylation site of Thr159 (amino acids 126-175)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising phospho peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to SRF (phospho Thr159) for IHC and ELISA with samples derived from Human and Mouse.
- Validated applications: IHC, ELISA
- Recommended dilutions: IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: Serum Response Factor SRF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse