Anti-FOXL2 Mouse Monoclonal Antibody [clone: PCRP-FOXL2-1B4]
Catalog # ANTIA314973-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Blepharophimosis
- Antigen symbol:FOXL2
- Clonality:Monoclonal
- Clone:PCRP-FOXL2-1B4
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2c
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P58012
- Antigen synonyms:Forkhead box protein L2|BPES1|BPES 1|BPES|FOX L2|Blepharophimosis epicanthus inversus and ptosis 1|Forkhead transcription factor FOXL2|Blepharophimosis epicanthus inversus and ptosis|Forkhead box L2
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:38 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Protein domain of FOXL2
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [PCRP-FOXL2-1B4] antibody to FOXL2 for ELISA and flow cytometry with samples derived from human.
- Validated applications: ELISA, flow cytometry
- Recommended dilutions: flow cytometry: 1 to 2 µg/million cells
Type: Primary
Antigen: FOXL2
Clonality: Monoclonal
Clone: PCRP-FOXL2-1B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2c
Reactivity: Human