Anti-CD23 Rabbit Recombinant Antibody [clone: FCER2/8511R]
Catalog # ANTIA314634-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:BLAST 2
- Antigen symbol:CD23
- Clonality:Recombinant
- Clone:FCER2/8511R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P06734
- Antigen synonyms:C-type lectin domain family 4 member J|C-type lectin domain family 4, member J|BLAST-2|C type lectin domain family 4 member J|CD23|CD 23A|CD 23|Blast2|CD23 antigen
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:37 kDa (soluble form)/45 kDa (membrane-bound form)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 221-321) of human FCER2/CD23 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [FCER2/8511R] antibody to CD23 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: CD23
Clonality: Recombinant
Clone: FCER2/8511R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human