Anti-HAND1 Rabbit Polyclonal Antibody
ANTIA97495-100
New Product
- Antibody type:Primary
- Antigen name:autonomic nervous system and neural crest derivatives-expressed protein 1
- Antigen symbol:HAND1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# O96004
- Antigen synonyms:Basic helix loop helix transcription factor HAND1|Extraembryonic tissues|Extraembryonic tissues heart autonomic nervous system and neural crest derivatives expressed protein 1|heart|HAND1_HUMAN|eHAND|HAND 1|bHLHa27|Class A basic helix-loop-helix protein 27
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:23 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human HAND1 (amino acids 141-190)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to HAND1 for IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:10000
Type: Primary
Antigen: HAND1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat