Anti-CD63 Mouse Monoclonal Antibody [Clone: MEM-259] (APC (Allophycocyanin))
Catalog # ANTIA85968-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD 63
- Antigen symbol:CD63
- Clonality:Monoclonal
- Clone:MEM-259
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P08962
- Antigen synonyms:CD63_HUMAN|gp55|CD63|CD63 molecule|Granulophysin|CD63 antigen (melanoma 1 antigen)|CD63 antigen|LAMP 3|CD63 antigen melanoma 1 antigen
- Storage buffer:Supplied in phosphate buffered saline with 15 mM sodium azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:HPB-ALL T cell line
- Purification:This antibody is conjugated with APC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [MEM-259] antibody to CD63 (APC) for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD63
Clonality: Monoclonal
Clone: MEM-259
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human