Order Entry
France
ContactUsLinkComponent
Anti-CBS Rabbit Monoclonal Antibody [clone: ARC0643]
Anti-CBS Rabbit Monoclonal Antibody [clone: ARC0643]
  ANTIA80837-100
New Product
undefined
Anti-CBS Rabbit Monoclonal Antibody [clone: ARC0643]
  ANTIA80837-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    AI047524
  • Antigen symbol:
    CBS
  • Clonality:
    Monoclonal
  • Clone:
    ARC0643
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P35520
  • Antigen synonyms:
    CBS|HIP 4|Beta-thionase|CBS_HUMAN|Cbs cystathionine beta-synthase|Beta thionase|Cystathionine beta synthase|AI303044|EC 4.2.1.22
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% Glycerol, 0,05% BSA, and 0,02% Sodium azide
  • Molecular weight:
    61 kDa
  • Sequence:
    EDLTEKKPWWWHLRVQELGLSAPLTVLPTITCGHTIEILREKGFDQAPVVDEAGVILGMVTLGNMLSSLLAGKVQPSDQVGKVIYKQFKQIRLTDTLGRLSHILEMDHFALVVHEQIQYHSTGKSSQRQMVFGVVTAIDLLNFVAAQERDQK
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 400-551 of human CBS (P35520)
  • Tested applications:
    IHC, ICC/IF
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit monoclonal [ARC0643] antibody to CBS for WB, IHC and ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF
  • Recommended dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:20

Type: Primary
Antigen: CBS
Clonality: Monoclonal
Clone: ARC0643
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat