Anti-CREB Goat Polyclonal Antibody
ANTIA82866-100
New Product
- Antibody type:Primary
- Antigen name:Active transcription factor CREB
- Antigen symbol:CREB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:cAMP response element binding protein 1|MGC9284|cAMP-responsive element-binding protein 1|cAMP responsive element binding protein 1|CREB-1|CREB1|cAMP response element binding protein|CREB1_HUMAN|Cyclic AMP-responsive element-binding protein 1
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% Sodium azide and 0,5% BSA
- Sequence:C-DSQESVDSVTDSQK
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human CREB1 (internal region)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Goat polyclonal antibody to CREB for ELISA, WB and IHC with samples derived from human and mouse.
- Validated applications: ELISA, WB, IHC
Type: Primary
Antigen: CREB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse