Anti-MXI1 Mouse Monoclonal Antibody [clone: PCRP-MXI1-1A3]
Catalog # ANTIA249446-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:bHLHc11
- Antigen symbol:MXI1
- Clonality:Monoclonal
- Clone:PCRP-MXI1-1A3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P50539
- Antigen synonyms:MAX-interacting protein 1|MAD 2|MGC43220|Class C basic helix-loop-helix protein 11|MAX dimerization protein 2|Max interactor 1|MAD2|MAX interacting protein 1|Max related transcription factor
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:26 kDa/22 kDa/33 kDa/21 kDa (isoforms)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human MXI1 protein
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [PCRP-MXI1-1A3] antibody to MXI1 for flow cytometry, IF and WB with samples derived from human.
- Validated applications: Flow cytometry, IF, WB
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml
Type: Primary
Antigen: MXI1
Clonality: Monoclonal
Clone: PCRP-MXI1-1A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human