Order Entry
France
ContactUsLinkComponent
Anti-CD23 Rabbit Recombinant Antibody [clone: FCER2/4395R]
Anti-CD23 Rabbit Recombinant Antibody [clone: FCER2/4395R]
  ANTIA248530-100
New Product
undefined
Anti-CD23 Rabbit Recombinant Antibody [clone: FCER2/4395R]
  ANTIA248530-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    BLAST 2
  • Antigen symbol:
    CD23
  • Clonality:
    Recombinant
  • Clone:
    FCER2/4395R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P06734
  • Antigen synonyms:
    C-type lectin domain family 4 member J|C-type lectin domain family 4, member J|BLAST-2|C type lectin domain family 4 member J|CD23|CD 23A|CD 23|Blast2|CD23 antigen
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    37 kDa (soluble form)/45 kDa (membrane - bound form)
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 221-321, of human CD23 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant rabbit monoclonal [FCER2/4395R] antibody to CD23 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CD23
Clonality: Recombinant
Clone: FCER2/4395R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human