Order Entry
France
ContactUsLinkComponent
Anti-ELK1 Mouse Monoclonal Antibody [clone: PCRP-ELK1-1B9]
Anti-ELK1 Mouse Monoclonal Antibody [clone: PCRP-ELK1-1B9]
Catalog # ANTIA248420-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-ELK1 Mouse Monoclonal Antibody [clone: PCRP-ELK1-1B9]
Catalog # ANTIA248420-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ELK 1
  • Antigen symbol:
    ELK1
  • Clonality:
    Monoclonal
  • Clone:
    PCRP-ELK1-1B9
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P19419
  • Antigen synonyms:
    ETS domain containing protein Elk1|ELK1|ELK1_HUMAN|ELK1, ETS transcription factor|ELK1 protein|ELK2 member of ETS oncogene family|ELK1 member of ETS oncogene family|ETS domain containing protein Elk 1|ETS domain protein Elk1
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    44,89 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human ELK1 protein
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [PCRP-ELK1-1B9] antibody to ELK1 for IP with samples derived from human.

  • Validated applications: IP
  • Recommended dilutions: IP: 1 to 2 µg/100 to 500 µg protein

Type: Primary
Antigen: ELK1
Clonality: Monoclonal
Clone: PCRP-ELK1-1B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human