(EZ)-Methacrifos 100 µg/ml in Acetonitrile
Catalog # EHERDREA14970100AL
Supplier: EHRENSTORFER
New Product
CAS Number:
Specifications
- Pk:1
- CAS No.:30864-28-9
- Description:(EZ)-Methacrifos 100 µg/ml in acetonitrile
- Shipping conditions:20 °C
- Synonym(s):Acrylic acid, 3-hydroxy-2-methyl-, methyl ester, O-ester with O,O-dimethyl phosphorothioate (8CI), Damfin, Metacriphos, Methacrifos, Methacriphos, Methyl 3-[(dimethoxyphosphinothioyl)oxy]-2-methyl-2-propenoate, O-[2-(Methoxycarbonyl)propenyl] O,O-dimethyl phosphorothioate, 2-Propenoic acid, 3-[(dimethoxyphosphinothioyl)oxy]-2-methyl-, methyl ester, (EZ)-Methacrifos
- Formula:C₇H₁₃O₅PS
- Storage temperature:–18 °C
- Molecular weight (Mw):240,21