Order Entry
France
Orders LinkContactUsLinkComponent
Mouse MG53 ELISA Kit

Mouse MG53 ELISA Kit

Mouse MG53 ELISA kit, 96 Tests
Catalog # ANTIA75922-96
Supplier:   Antibodies.com
Mouse MG53 ELISA Kit
Catalog # ANTIA75922-96
Supplier:  Antibodies.com
CAS Number:  
Mouse MG53 ELISA Kit
Mouse MG53 ELISA Kit

Mouse MG53 ELISA kit, 96 Tests

CAS Number:  
Price
625,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Mouse MG53 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse MG53 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 78,125 to 5000 pg/ml
  • Sensitivity: 46,875 pg/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse MG53 ELISA kit (A75922) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse MG53 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for MG53 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the MG53 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-MG53 Antibody, which binds the captured MG53 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of MG53 captured in each well. The concentration of MG53 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Pk:
    96 Tests
  • Description:
    Mouse MG53 ELISA kit
  • Primary antibody reactivity:
    Mouse
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Sample volume:
    100 μl
  • Detection range:
    78,125 - 5000 pg/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Cross reactivity:
    Mouse MG53 ELISA kit exhibits high specificity and excellent specificity for the detection of mouse MG53. No significant cross-reactivity or interference between MG53 and analogues was observed.
  • Assay Principle:
    Quantitative
  • Cat. No.:
    ANTIA75922-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    46,875 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    MG53
  • Regulatory status:
    RUO

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.