Anti-CD138 Mouse Monoclonal Antibody [clone: 1A3H4]
Catalog # NOVUNBP2-37282
Supplier: Novus Biologicals
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD138 antigen
- Antigen symbol:CD138
- Clonality:Monoclonal
- Clone:1A3H4
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western blot:Yes
- Format:BSA Free
- Gene ID:SDC1
- Antigen synonyms:syndecan 1|SDC|Syndecan-1|syndecan proteoglycan 1|SYND1heparan sulfate proteoglycan fibroblast growth factor receptor|syndecan|SDC1|Syndecan1
- Purity:Protein G purified
- Storage buffer:PBS
- Storage temperature:Store at 4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Purified recombinant fragment of human Syndecan-1/CD138 (AA: 28-171) expressed in E. Coli
- Pk:0,1 mL
Specifications
About this item
This antibody is CyTOF-ready.
- ELISA 1:10000
- Flow Cytometry 1:200 - 1:400
- Immunocytochemistry/ Immunofluorescence 1:200 - 1:1000
- Immunohistochemistry 1:200 - 1:1000
- Immunohistochemistry-Paraffin 1:200 - 1:1000
- Western Blot 1:500 - 1:2000
this antibody is reactive to Mouse and derived from the same host, Mouse. Mouse-On-Mouse blocking reagent may be needed for IHC and ICC experiments to reduce high background signal.
Caution: This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Type: Primary
Antigen: CD138
Clonality: Monoclonal
Clone: 1A3H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse