Order Entry
France
ContactUsLinkComponent
 
Human Recombinant IGF-I, ACF
 
undefined
Human Recombinant IGF-I, ACF

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein synonyms:
    IBP1|IGF-IA|IGF-IB|IGF1A|Insulin-like growth factor 1|Mechano growth factor|MGF|Somatomedin C
  • Protein/peptide name:
    IGF-I, ACF
  • Purity:
    ≥95%

 

Frequently Bought Together