Order Entry
France
ContactUsLinkComponent
Human Recombinant PDGF-AB
Human Recombinant PDGF-AB
Certificates
undefined
Human Recombinant PDGF-AB

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    PDGF-AB
  • Purity:
    ≥95%

Specifications

Frequently Bought Together