Order Entry
France
ContactUsLinkComponent
EBV (HLA Class I Control) Peptide Pool
EBV (HLA Class I Control) Peptide Pool
Catalog # STMC100-1391
CAS Number:  
undefined
EBV (HLA Class I Control) Peptide Pool
Catalog # STMC100-1391
CAS Number:  

Specifications

  • Pk:
    1 AU
  • Seq:
    0001
  • Protein/peptide name:
    EBV (HLA Class I Control) Peptide Pool
  • Purity:
    ≥95%

Specifications

About this item

Stimulating T cells with EBV (HLA Class I Control) peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. EBV (HLA Class I Control) Peptide Pool is a lyophilised mixture of 26 peptides from Epstein-Barr virus (EBV), and consists of defined HLA class I-restricted T cell epitopes. Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development. For consistency and reproducibility across your applications, CMV peptide pool from STEMCELL has a purity ≥95%.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').