Order Entry
France
ContactUsLinkComponent
SARS-CoV-2 (Spike Protein) Delta/B.1.617.2 WT Reference Peptide Pool
SARS-CoV-2 (Spike Protein) Delta/B.1.617.2 WT Reference Peptide Pool
  STMC100-1381
 :  
undefined
SARS-CoV-2 (Spike Protein) Delta/B.1.617.2 WT Reference Peptide Pool
  STMC100-1381
 :  
  • Pk:
    1 AU
  • Seq:
    0001
  • Protein/peptide name:
    SARS-CoV-2 (Spike Protein) Delta/B.1.617.2 WT Reference Peptide Pool
  • Purity:
    Average 70%
Stimulating T cells with SARS-CoV-2 (Spike Protein) Delta/B.1.617.2 WT reference peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. This peptide pool is a lyophilised mixture of 27 peptides from the spike glycoprotein of the wild-type SARS-CoV-2 virus. Consisting of the wild-type peptides that correspond to the mutated portions of the spike protein found in the SARS-CoV-2 Delta variant (B.1.617.2), this pool is recommended to be used as a reference for the SARS-CoV-2 (Spike Protein) Delta/B.1.617.2 Mutation Peptide Pool. Through interactions with the angiotensin-converting enzyme 2 (ACE2) receptor, the spike protein enables the virus to attach to the cell membrane, and plays a critical role in viral entry (Hoffmann et al.; Walls et al.). Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').