Order Entry
France
ContactUsLinkComponent
 
Mouse Recombinant IGF-I
 
undefined
Mouse Recombinant IGF-I

 

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Protein/peptide name:
    IGF-I
  • Purity:
    ≥95%

 

Frequently Bought Together