Order Entry
France
ContactUsLinkComponent
Rhodamine 19 perchlorate ~95% (dye content), Sigma-Aldrich®
Rhodamine 19 perchlorate ~95% (dye content), Sigma-Aldrich®
  546062-100MG-A
 :  
undefined
Rhodamine 19 perchlorate ~95% (dye content), Sigma-Aldrich®
  546062-100MG-A
 :  

 

  • Pk:
    100 mg
  • Seq:
    2440

 

 

  • Appearance (colour):
    Brown to very dark brown and red to very dark red and red-brown and very dark red-brown and green to very dark green
  • Appearance (form):
    Powder or crystals
  • Purity (hplc area %):
    ~95%
  • Loss on drying:
    ≤10%
  • Carbon content:
    60.64% (theory)
  • Nitrogen content:
    5.44% (theory)
  • Wavelength (1) (UV):
    512 - 522 nm
  • Molar absorbancy index (1):
    Report result
  • Wavelength (2) (UV):
    346 - 356 nm
  • Molar absorbancy index (2):
    Report result
  • Wavelength (3) (UV):
    289 - 299 nm
  • Molar absorbancy index (3):
    Report result
  • Wavelength (4) (UV):
    265 - 275 nm
  • Molar absorbancy index (4):
    Report result
  • Wavelength (5) (UV):
    240 - 250 nm
  • Molar absorbancy index (5):
    Report result
  • Solvent (UV):
    Ethanol

 

 

 :
 :514.96 g/mol
 :>300 °C (lit.)
 :M-Clarity™ quality level = MQ100
 :Room Temperature
 :263-686-2
 :62669-66-3
 :C₂₆H₂₇ClN₂O₇
 :MFCD02684062