



Crystal violet, Sigma-Aldrich®
:548-62-9
:
:C₂₅H₃₀ClN₃
:MFCD00011750
:407.99 g/mol
:13,04406
:189…194 °C
:1,19 g/cm³ (20 °C)
:208-953-6
:3077
:9,III
:GHS05
:GHS08
:GHS07
:GHS09
:Room Temperature
- Appearance (colour):Green to very dark green
- Appearance (form):Powder
- Thin layer chromatogr.:Corresponds to standard
- Solubility (colour):0.01 g in 10 ml methanol
- Solubility (method):0.01 g in 10 ml methanol
- Loss on drying:≤10%
- Wavelength (1) (UV):580 - 595 nm
- Extinction (1%/1 cm) (1) (UV):≥1300
- Solvent (UV):Water
Frequently Bought Together