Crystal violet, certified
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
- CAS Number:548-62-9
- Formula:C₂₅H₃₀ClN₃
- MDL Number:MFCD00011750
- Molecular Weight:407.99 g/mol
- Merck Index:13,04406
- Melting Pt:173 °C (dec.)
- Density:1,19 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:208-953-6
- UN:3077
- ADR:9,III
3 Options Available Below
Product Details & Documents
Synonyms: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
- CAS Number:548-62-9
- Formula:C₂₅H₃₀ClN₃
- MDL Number:MFCD00011750
- Molecular Weight:407.99 g/mol
- Merck Index:13,04406
- Melting Pt:173 °C (dec.)
- Density:1,19 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:208-953-6
- UN:3077
- ADR:9,III
Documents
Specifications for Product Family
Specification Test Results
- Appearance:Dark green Crystalline powder
- Infrared spectrum:Conforms
- Dye content:≥88 %
- UV Lambda max:589 to 594 nm
- Absorption Ratio P-15/P+15:0.98 to 1.20
- BSC Certification:Passes
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



