Order Entry
France
ContactUsLinkComponent
Pesticide reference standard, Azinphos-methyl 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, Azinphos-methyl 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL10365000CY
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, Azinphos-methyl 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL10365000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    86-50-0
  • Description:
    Azinphos-methyl 10 µg/ml in cyclohexane
  • Formula:
    C₁₀H₁₂N₃O₃PS₂
  • Description:
    Azinphos-methyl

Specifications

About this item