Pesticide reference standard, Azinphos-ethyl 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA10360000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:2642-71-9
- Description:Azinphos-ethyl 100 µg/ml in cyclohexane
- Formula:C₁₂H₁₆N₃O₃PS₂
- Description:Azinphos-ethyl