Order Entry
France
Orders LinkContactUsLinkComponent
TeSR™-E6 Medium
TeSR™-E6 Medium
TeSR™ E6 serum and xeno-free medium, 1 KIT
Catalog # STMC05946
Marketsource
TeSR™-E6 Medium
Catalog # STMC05946
Marketsource
CAS Number:  
TeSR™-E6 Medium
TeSR™-E6 Medium
TeSR™ E6 serum and xeno-free medium, 1 KIT
CAS Number:  
Price
448,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Defined, serum-free, xeno-free medium for pluripotent stem cells.
TeSR™-E6 is a defined, serum- and xeno-free medium that is based on the formulation of TeSR™-E8™, but does not contain transforming growth factor β (TGF-β) or basic fibroblast growth factor (bFGF). It may be used as a basal medium for differentiation of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells, or other applications where removal of the above cytokines is desirable.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    TeSR™ E6 serum and xeno-free medium