Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (PE)
Supplier: AAT Bioquest
Certificates
About this item
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.
- Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Caution: For research use only.
Type: Primary
Antigen: CD14 antigen
Clonality: Monoclonal
Clone: 61D3
Conjugation: PE
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- AATB101411L2
- AATB101411L0
- AATB101411L1
- Antigen symbol:
- CD14
- CD14
- CD14
- Antigen name:
- CD14 antigen
- CD14 antigen
- CD14 antigen
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- 61D3
- 61D3
- 61D3
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Yes
- Purification:
- Affinity chromatography
- Affinity chromatography
- Affinity chromatography
- Cross adsorption:
- No
- No
- No
- Storage temperature:
- 2...8 °C with minimised light exposure. Do not freeze.
- 2...8 °C with minimised light exposure. Do not freeze.
- 2...8 °C with minimised light exposure. Do not freeze.
- Shipping temperature:
- 20
- 20
- 20
Specifications
Frequently Bought Together