Anti-BTNL3 Rabbit Polyclonal Antibody
PRSI55-499
New Product
- Antibody type:Primary
- Antigen name:Butyrophilin-like protein 3
- Antigen symbol:BTNL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Western blot:Yes
- Epitope:Amino Acid Range = 427-455
- Form:Liquid
- Gene ID:Q6UXE8
- Antigen synonyms:BTNL3
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:52 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This BTNL3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 427-455 amino acids from the C-terminal region of human BTNL3.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Size:400 µl
- Pk:400 µl
BTNL3 is expressed in small intestine, colon, testis, spleen, and leukocyte and plays a role in lipid metabolism. Two isoforms have been described.
For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: BTNL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human