Anti-ATL3 Rabbit Polyclonal Antibody
PRSI14-307
New Product
- Antibody type:Primary
- Antigen name:atlastin GTPase 3
- Antigen symbol:ATL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:25923
- Antigen synonyms:HSN1F|ATL3|DKFZP564J0863
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 360-440 of human ATL3 (NP_056274.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: ATL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat